PRIH93ED7DD8 Princeton BioMolecular Research, Inc.

5-(2-Methoxy-phenoxymethyl)-4-phenyl-4H-[1,2,4]triazole-3-thiol

  • MDL number MFCD01809911
  • Molecular Weight : 313.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart