PRIH93ED7DE6 Princeton BioMolecular Research, Inc.

1-{4-[1-(2-Hydroxy-ethyl)-1H-benzoimidazol-2-ylmethoxy]-phenyl}-ethanone

  • MDL number MFCD06755453
  • Cas Number : 891449-68-6
  • Molecular Weight : 310.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart