PRIH93ED7DF3 Princeton BioMolecular Research, Inc.

2-[2-(2-Trifluoromethyl-benzoimidazol-1-ylmethyl)-benzoimidazol-1-yl]-ethanol

  • MDL number MFCD06755499
  • Molecular Weight : 360.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart