PRIH93ED7DF4 Princeton BioMolecular Research, Inc.

2-Ureidomethylene-malonic acid diethyl ester

  • MDL number MFCD00193763
  • Cas Number : 61679-84-3
  • Molecular Weight : 230.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart