PRIH93ED7E14 Princeton BioMolecular Research, Inc.

1-Allyl-3-naphthalen-1-yl-thiourea

  • MDL number MFCD00091308
  • Cas Number : 17073-29-9
  • Molecular Weight : 242.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart