PRIH93ED7E15 Princeton BioMolecular Research, Inc.

1-Allyl-3-cyclohexyl-thiourea

  • MDL number MFCD00466091
  • Cas Number : 77563-68-9
  • Molecular Weight : 198.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart