PRIH93ED7E16 Princeton BioMolecular Research, Inc.

3-(6-Fluoro-4-oxo-4H-quinolin-1-yl)-propionic acid

  • MDL number MFCD18784696
  • Cas Number : 1267499-14-8
  • Molecular Weight : 235.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart