PRIH93ED7E1D Princeton BioMolecular Research, Inc.

(2-Trifluoromethyl-furan-3-yl)-acetic acid

  • MDL number MFCD19981832
  • Molecular Weight : 194.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart