PRIH93ED7E2B Princeton BioMolecular Research, Inc.

8-Fluoro-2-methyl-5,7-dinitro-quinoline

  • MDL number MFCD19981931
  • Molecular Weight : 251.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart