PRIH93ED7E48 Princeton BioMolecular Research, Inc.

3-Nitro-dibenzo[b,f]oxepin-1-ylamine

  • MDL number MFCD02612727
  • Cas Number : 361374-55-2
  • Molecular Weight : 254.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart