PRIH93ED7E6E Princeton BioMolecular Research, Inc.

1H,1'H-[2,5']Bibenzoimidazolyl-2'-thiol

  • MDL number MFCD21601773
  • Molecular Weight : 266.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart