PRIH93ED7E74 Princeton BioMolecular Research, Inc.

4-m-Tolyl-morpholine

  • MDL number MFCD05854326
  • Cas Number : 7025-91-4
  • Molecular Weight : 177.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart