PRIH93ED7E7E Princeton BioMolecular Research, Inc.

2-Bromo-4,7,7-trimethyl-3-oxo-bicyclo[2.2.1]heptane-1-carboxylic acid methylamide

  • MDL number MFCD04088751
  • Molecular Weight : 288.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart