PRIH93ED7E82 Princeton BioMolecular Research, Inc.

3-Chloromethyl-2-trifluoromethyl-furan

  • MDL number MFCD22371776
  • Molecular Weight : 184.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart