PRIH93ED7E92 Princeton BioMolecular Research, Inc.

6-Amino-9-butyl-9H-purine-8-thiol

  • MDL number MFCD09864092
  • Cas Number : 521308-65-6
  • Molecular Weight : 223.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart