PRIH93ED7E9F Princeton BioMolecular Research, Inc.

2-(2-Mercapto-benzoimidazol-1-yl)-acetamide

  • MDL number MFCD11201516
  • Molecular Weight : 207.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart