PRIH93ED7EA4 Princeton BioMolecular Research, Inc.

4-Bromo-1,7,7-trimethyl-bicyclo[2.2.1]heptan-2-one oxime

  • MDL number MFCD22688591
  • Molecular Weight : 246.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart