PRIH93ED7EA6 Princeton BioMolecular Research, Inc.

1,4,7,7-Tetramethyl-bicyclo[2.2.1]heptane-2,3-dione

  • MDL number MFCD06291733
  • Cas Number : 62292-65-3
  • Molecular Weight : 180.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart