PRIH93ED7EB3 Princeton BioMolecular Research, Inc.

1-Phenyl-3-(1-phenyl-ethyl)-thiourea

  • MDL number MFCD00158843
  • Cas Number : 15093-41-1
  • Molecular Weight : 256.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart