PRIH93ED7EBC Princeton BioMolecular Research, Inc.

7-Ethylsulfanyl-3-phenyl-[1,2,4]triazolo[5,1-c][1,2,4]triazin-4-ylamine

  • MDL number MFCD26098500
  • Cas Number : 1356830-91-5
  • Molecular Weight : 272.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart