PRIH93ED7EC0 Princeton BioMolecular Research, Inc.

3-Methyl-6-nitro-1H-quinoxalin-2-one

  • MDL number MFCD26130604
  • Cas Number : 19801-10-6
  • Molecular Weight : 205.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart