PRIH93ED7ECA Princeton BioMolecular Research, Inc.

4-(4,5-Dimethyl-2-oxo-2,3-dihydro-imidazol-1-yl)-benzoic acid

  • MDL number MFCD26130612
  • Cas Number : 31409-39-9
  • Molecular Weight : 232.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart