PRIH93ED7EF5 Princeton BioMolecular Research, Inc.

2-(3,5-Dimethyl-isoxazole-4-sulfonylamino)-benzoic acid

  • MDL number MFCD13563193
  • Cas Number : 1042769-63-0
  • Molecular Weight : 296.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart