PRIH93ED7EF7 Princeton BioMolecular Research, Inc.

2-(3,5-Dimethyl-isoxazole-4-sulfonylamino)-4-methyl-pentanoic acid

  • MDL number MFCD09037198
  • Molecular Weight : 290.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart