PRIH93ED7F0D Princeton BioMolecular Research, Inc.

2-Amino-5-trifluoromethyl-[1,2,4]triazolo[1,5-a]pyrimidin-7-ol

  • MDL number MFCD01360116
  • Cas Number : 326906-13-2
  • Molecular Weight : 219.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart