PRIH93ED7F11 Princeton BioMolecular Research, Inc.

(2-Amino-6-oxo-thiazolo[3,2-b][1,2,4]triazol-5-ylidene)-acetic acid methyl ester

  • MDL number MFCD03545908
  • Molecular Weight : 226.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart