PRIH93ED7F1D Princeton BioMolecular Research, Inc.

(3-Amino-phenyl)-thiophen-2-yl-methanone

  • MDL number MFCD09035567
  • Molecular Weight : 203.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart