PRIH93ED7F35 Princeton BioMolecular Research, Inc.

4-[1-(4-Methoxy-phenyl)-1H-tetrazol-5-ylmethoxy]-benzoic acid

  • MDL number MFCD09761251
  • Molecular Weight : 326.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart