PRIH93ED7F3C Princeton BioMolecular Research, Inc.

4-[5-(4-Bromo-phenyl)-3-(6-methyl-2-oxo-4-phenyl-1,2-dihydro-quinolin-3-yl)-4,5-dihydro-pyrazol-1-yl]-4-oxo-butyric acid

  • MDL number MFCD02989065
  • Cas Number : 380560-89-4
  • Molecular Weight : 558.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart