PRIH93ED7F41 Princeton BioMolecular Research, Inc.

N,N'-Bis-(4-methyl-phthalazin-1-yl)-hydrazine

  • MDL number MFCD26097368
  • Molecular Weight : 316.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart