PRIH93ED7F46 Princeton BioMolecular Research, Inc.

Bis-(1-methyl-heptyl)-amine

  • MDL number MFCD00048901
  • Cas Number : 5412-92-0
  • Molecular Weight : 241.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart