PRIH93ED7F4B Princeton BioMolecular Research, Inc.

1-Thiazol-2-yl-pyrrolidin-2-one

  • MDL number MFCD00664608
  • Cas Number : 37762-96-2
  • Molecular Weight : 168.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart