PRIH93ED7F68 Princeton BioMolecular Research, Inc.

(9-Benzyl-9H-benzo[4,5]imidazo[2,1-c][1,2,4]triazol-3-ylsulfanyl)-acetic acid

  • MDL number MFCD04206118
  • Cas Number : 606120-73-4
  • Molecular Weight : 338.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart