PRIH93ED7F6B Princeton BioMolecular Research, Inc.

(3-Isopropyl-4-oxo-2-thioxo-thiazolidin-5-yl)-acetic acid

  • MDL number MFCD02258185
  • Cas Number : 56879-41-5
  • Molecular Weight : 233.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart