PRIH93ED7F77 Princeton BioMolecular Research, Inc.

3-Methyl-thiazolo[5,4-d]isoxazol-5-ylamine

  • MDL number MFCD28353694
  • Cas Number : 1706464-22-3
  • Molecular Weight : 155.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart