PRIH93ED7F83 Princeton BioMolecular Research, Inc.

(1Z,2Z)-N-hydroxy-2-hydroxyimino-2-phenyl-acetimidoyl chloride

  • MDL number MFCD28353701
  • Molecular Weight : 198.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart