PRIH93ED7F85 Princeton BioMolecular Research, Inc.

4-Hydroxy-3-methoxy-5-piperidin-1-ylmethyl-benzaldehyde

  • MDL number MFCD28353702
  • Cas Number : 79089-46-6
  • Molecular Weight : 249.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart