PRIH93ED7F90 Princeton BioMolecular Research, Inc.

2,5-Dimethyl-4-(1-methyl-1,2,3,6-tetrahydro-pyridin-4-yl)-2H-pyrazol-3-ylamine

  • MDL number MFCD24623997
  • Molecular Weight : 206.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart