PRIH93ED7FA4 Princeton BioMolecular Research, Inc.

[1-(2-Phenoxy-ethyl)-1H-benzoimidazol-2-yl]-phenyl-amine

  • MDL number MFCD03828056
  • Molecular Weight : 329.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart