PRIH93ED7FA7 Princeton BioMolecular Research, Inc.

2-[2-(4-tert-Butyl-phenoxy)-ethylsulfanyl]-5-methyl-1H-benzoimidazole

  • MDL number MFCD28398664
  • Molecular Weight : 340.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart