PRIH93ED7FB5 Princeton BioMolecular Research, Inc.

2-[2-(4-tert-Butyl-phenoxy)-ethylsulfanyl]-1-ethyl-1H-benzoimidazole

  • MDL number MFCD02647543
  • Molecular Weight : 354.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart