PRIH93ED7FC9 Princeton BioMolecular Research, Inc.

[9-(2-Phenoxy-ethyl)-9H-benzo[4,5]imidazo[2,1-c][1,2,4]triazol-3-ylsulfanyl]-acetic acid

  • MDL number MFCD03698284
  • Cas Number : 676630-78-7
  • Molecular Weight : 368.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart