PRIH93ED7FCC Princeton BioMolecular Research, Inc.

2-(Benzo[4,5]thiazolo[2,3-c][1,2,4]triazol-3-ylsulfanyl)-butyric acid

  • MDL number MFCD03765603
  • Molecular Weight : 293.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart