PRIH93ED7FD6 Princeton BioMolecular Research, Inc.

3-(2-p-Tolyloxy-ethylsulfanyl)-benzo[4,5]thiazolo[2,3-c][1,2,4]triazole

  • MDL number MFCD05271733
  • Molecular Weight : 341.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart