PRIH93ED7FD9 Princeton BioMolecular Research, Inc.

2-o-Tolyl-2,3-dihydro-isoindol-1-one

  • MDL number MFCD01014221
  • Cas Number : 17372-63-3
  • Molecular Weight : 223.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart