PRIH93ED7FFB Princeton BioMolecular Research, Inc.

2-Cyano-3-mercapto-3-methylsulfanyl-acrylamide

  • MDL number MFCD00119424
  • Cas Number : 111479-66-4
  • Molecular Weight : 174.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart