PRIH93ED7FFF Princeton BioMolecular Research, Inc.

1-(3-Formyl-4-hydroxy-5-methoxy-benzyl)-piperidine-4-carboxylic acid

  • MDL number MFCD29047733
  • Molecular Weight : 293.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart