PRIH93ED8047 Princeton BioMolecular Research, Inc.

2-(chloromethyl)-5-(2-naphthyl)-1,3,4-oxadiazole

  • MDL number MFCD01313793
  • Cas Number : 300665-29-6
  • Molecular Weight : 244.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart