PRIH93ED805A Princeton BioMolecular Research, Inc.

(2E)-2-[(aminocarbonothioyl)hydrazono]-N-ethylpropanamide

  • MDL number MFCD01361007
  • Molecular Weight : 188.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart