PRIH93ED808A Princeton BioMolecular Research, Inc.

4-[(dimethylamino)methyl]-N-(1-methyl-2-phenylethyl)benzamide

  • MDL number MFCD30744939
  • Molecular Weight : 296.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart